Learn More
Abcam Agomelatine, MT1/MT2 agonist and 5-HT2C antagonist, 100MG
Supplier:  Abcam AB142476100MG
Encompass_Preferred
MW 243.3 Da, Purity >99%. Potent MT1/MT2 agonist (Ki values are 0.1 and 0.12 nM, respectively). 5-HT2C antagonist (IC50 = 270 nM). Shows antidepressant effects *in vivo*. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6230