Learn More
Medchemexpress LLC Agrimol B | 55576-66-4 | 682.75 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN07045MG
Encompass_Preferred
Agrimol B is a polyphenol that acts as an orally active and potent SIRT1 activator. It demonstrates anti-adipogenic, anticancer, and antibacterial activity against plant pathogens. It significantly inhibits 3T3-L1 adipocyte differentiation by reducing the expression of PPARγ, C/EBPα, FAS, UCP-1, and apoE. Its effect on cancer cells is believed to stem from its influence on c-MYC, SKP2, and p27.
  • Polyphenol
  • Orally active and potent SIRT1 activator
  • Anti-adipogenic activity
  • Anticancer activity
  • Antibacterial activity against plant pathogens
  • Inhibits 3T3-L1 adipocyte differentiation
  • Affects c-MYC, SKP2, and p27 in cancer cells
Catalog No. 50-004-15400