Learn More
Medchemexpress LLC Agrimol B | 55576-66-4 | MFCD20527284 | 99.5% | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN070410MG
Encompass_Preferred
Agrimol B is a polyphenolic natural product that acts as an orally active, potent SIRT1 activator with reported anti-adipogenic, anticancer, and antibacterial activities. It modulates pathways involved in adipocyte differentiation and cell-cycle regulation, making it useful for research in metabolic and cancer biology.
  • Potent SIRT1 activator with oral activity.
  • Exhibits anti-adipogenic activity by inhibiting adipocyte differentiation.
  • Shows anticancer effects linked to modulation of c-MYC, SKP2, and p27.
  • High purity (>99%) solid, white to yellow in appearance.
  • Soluble in DMSO at approximately 5 mg/mL with sonication and heating.
  • Stable as powder at -20°C for years; store in solvent at -80°C for months.
Catalog No. 50-002-95941