Learn More
Medchemexpress LLC AGRP protein, mouse | ≥95.0% | 100 UG

Supplier: Medchemexpress LLC HYP75524100UG
Recombinant mouse agouti-related protein (C-terminal 50 amino acids) expressed in HEK293 as an Fc fusion and supplied as a purified protein fragment for research into melanocortin receptor signaling, appetite regulation, and metabolic studies. The material includes quality controls for endotoxin and has recommended storage and handling to preserve activity.
- 50 amino acid C-terminal fragment suitable for receptor binding studies.
- Expressed in HEK293 cells to support mammalian post-translational modifications.
- Fc fusion tag for improved stability and detection.
- Purity ≥95.0% as determined by reducing SDS-PAGE.
- Endotoxin level <1 EU/μg to minimize assay interference.
- Store at -20°C; after reconstitution stable at 4°C for 1 week or -20°C for longer.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.