Learn More
Medchemexpress LLC AGS-67E 10mg | 10mg
SDP
Supplier:  Medchemexpress LLC HYP99031710MG
Encompass_Preferred
AGS-67E is a humanized antibody expressed by CHO targeting TSPAN26/CD37 AGS-67E has a huIgG1 heavy chain and a hu light chain with a predicted molecular weight (MW) of 143 44 kDa The isotype control for AGS-67E can be referenced as Human IgG2 kappa Isotype Control (HY-P99002)
Catalog No. 50-003-25596