Learn More
ABclonal Technology AGTRAP Rabbit pAb
SDP
Supplier:  ABclonal Technology A12178100UL
Encompass_Preferred
This gene encodes a transmembrane protein localized to the plasma membrane and perinuclear vesicular structures. The gene product interacts with the angiotensin II type I receptor and negatively regulates angiotensin II signaling. Alternative splicing of this gene generates multiple transcript variants encoding different isoforms.
Catalog No. 50-001-16373