Learn More
Medchemexpress LLC AGX51 | 330834-54-3 | 99.3% | 431.52 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY129241
Encompass_Preferred
AGX51 is a first-in-class pan-Id (inhibitors of DNA-binding/differentiation proteins) antagonist and degrader, primarily used in cancer research. It functions by inhibiting Id1-E47 interaction, leading to ubiquitin-mediated degradation of Ids. This action results in cell growth arrest and a reduction in cell viability, demonstrating an ability to inhibit Triple-Negative Breast Cancer (TNBC) with an IC50 of approximately 25 nM.
  • First-in-class pan-Id antagonist and degrader
  • Inhibits Id1-E47 interaction
  • Leads to ubiquitin-mediated degradation of Ids
  • Results in cell growth arrest and viability reduction
  • Effective in inhibiting Triple-Negative Breast Cancer (TNBC)
Catalog No. 50-003-64293