Learn More
Enzo Life Sciences AH-6809 (5 mg). CAS: 33458-93-4
SDP
Supplier:  Enzo Life Sciences BMLRA1100005
Encompass_Preferred
Selective prostaglandin E2 receptor (EP1) antagonist. Useful at concentrations up to 10 µM, at which point it starts to block PGD2 and TXA2 receptors. Purity: ≥98% (TLC). Solubility: Soluble in DMSO. Long Term Storage: Ambient.
Catalog No. 50-201-1714