Learn More
TRIBIOSCIENCE INC Adenosyl Homocysteinase (AHCY) Fluorometric Assay Kit, 100 Tests
SDP
Supplier:  TRIBIOSCIENCE INC TBS2056
Encompass_Preferred
AHCY Activity Fluorometric Assay provides the easiest and most accurate approach to measuring AHCY activity in a variety of samples. The free thiol group of the homocysteine can be detected with a thiol probe generating an enhanced fluorescent signal at Ex/Em = 400/465 nm.
Catalog No. NC2886181