Learn More
TRIBIOSCIENCE INC Adenosylhomocysteine (AHCY) Inhibitor Screening Assay Kit (Fluorometric), 100 Tests
SDP
Supplier:  TRIBIOSCIENCE INC TBS2099100
Encompass_Preferred
AHCY inhibitor screening assay provides the easiest and most accurate approach to measuring AHCY activity. The free thiol group of the homocysteine can be detected with a thiol probe generating an enhanced fluorescent signal at Ex/Em = 400/465 nm.
Catalog No. NC2887478