Learn More
TARGETMOL CHEMICALS INC AHR Inhibitor I-103 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T853431MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. AHR Inhibitor I-103 is an aryl hydrocarbon receptor (AHR) inhibitor with anticancer activity used in the study of breast cancer and acute myeloid leukemia. Purity 98.64%
Catalog No. 50-287-6040