Learn More
Selleck Chemical LLC AI-10-49
SDP
Supplier:  Selleck Chemical LLC S781425MG
Encompass_Preferred
AI-10-49 is a selective inhibitor of the binding of CBF -SMMHC to RUNX1 with IC50 of 260 nM
Catalog No. 50-000-04595