Learn More
Across International Ai 3x2" Pneumatic Heat Press with Dual Heating Platens - 110V
SDP
Supplier:  Across International AIRPRESS0302
Encompass_Preferred
The new 2016 Ai AirPress 3 x 2 compact pneumatic heat press features dual built-in digital thermometers and heaters, allowing you to precisely control the temperature of your transfer. The digital timer will automatically begin a countdown as soon as the unit is pressed shut and provides an audible alarm once the predetermined amount of time has elapsed.
Catalog No. 50-225-1307