Learn More
Medchemexpress LLC HY-13417 500mg , AICAR Acadesine;AICA Riboside CAS:2627-69-2 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY13417500MG
Encompass_Preferred
Medchemexpress, HY-13417 500mg AICAR Acadesine;AICA Riboside CAS:2627-69-2 Formula:C9H14N4O5 AMPK Purity:98% AICAR is an activator of AMP-activated protein kinase ( AMPK ), down-regulates the insulin receptor expression in HepG2 cells. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2361
 Restricted Item