Learn More
Enzo Life Sciences AICAR, (250mg), CAS Number: 2627-69-2
SDP
Supplier:  Enzo Life Sciences BMLEI3300250
Encompass_Preferred
AMPK activator. Alternative Name: 5-Aminoimidazole-4-carboxamide 1-beta-D-ribofuranoside, Acadesine, Formula: C9H14N4O5. MW: 258.2. CAS Number: 2627-69-2. Purity: ≥98% (TLC). Appearance: Off-white solid. Solubility: Soluble in water (9mg/ml). Long Term Storage: -20°C
Catalog No. NC9429088