Learn More
Selleck Chemical LLC AICAR (Acadesine) S1802-1g
SDP
Supplier:  Selleck Chemical LLC S18021G
Encompass_Preferred
AICAR (Acadesine NSC105823 AICA Riboside) an AMPK activator results in accumulation of ZMP which mimics the stimulating effect of AMP on AMPK and AMPK kinase AICAR (Acadesine) induces mitophagy Phase 3
Catalog No. 50-310-9137