Learn More
Cayman Chemical AICAR, 2627-69-2, 100 mg
SDP
Supplier:  Cayman Chemical 10010241100
Encompass_Preferred
Molecular Formula C9H14N4O5, Purity ≥98%, Formula Weight 258.2, Synonyms: 5-amino-1-β-D-ribofuranosyl-1H-imidazole-4-carboxamide, Acadesine, AICA-Riboside, NSC 105823; A selective activator of AMPK; inhibits synthesis of fatty acids and sterols in hepatocytes and insulin-stimulated glucose uptake in adipocytes.
Catalog No. NC9970786