Learn More
ABclonal Technology AIF1/IBA1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15271000UL
Encompass_Preferred
This gene encodes a protein that binds actin and calcium. This gene is induced by cytokines and interferon and may promote macrophage activation and growth of vascular smooth muscle cells and T-lymphocytes. Polymorphisms in this gene may be associated with systemic sclerosis. Alternative splicing results in multiple transcript variants, but the full-length and coding nature of some of these variants is not certain.
Catalog No. 50-001-06789