Learn More
MSC Rochester Midland Corporation Air Freshener, Liquid, 1 gal, Pine Scent, Concentrated
Supplier:  MSC 54472758
Encompass
Fresh Aire liquid air freshener is a concentrated, water-based product designed to neutralize chemical mal-odors and leave the air clean and fresh. Packaged in a 1 gal bottle with a refreshing pine scent, it can be dispensed by pour or spray. Ideal for use in locker rooms, restrooms, on fabric, carpet, for garbage, bathrooms, and trash.
  • Excellent fragrance booster
  • Highly concentrated formula
  • Extended product coverage
  • Cost effective solution
  • Fresh, clean fragrance
  • Clear green color
  • Neutral pH of 7.0
Catalog No. 50-283-7975