Learn More
Chemglass Life Sciences Chemglass Air Operated Lower Drain Valve for 10-50 L Reactors (CG-1968-V-01)
SDP
Supplier:  Chemglass Life Sciences CG1968V01
Encompass
Air-operated flush bottom drain valve designed for 10 L-50 L reactors with a 2 in beaded pipe connection. The valve is held closed by spring tension and opens when air pressure is applied. Supplied as a complete kit with an air control panel, two regulators, a mounting bracket, tubing, and fittings. Suitable for solvent extractions and controlled drain operations; not recommended for slurries or viscous materials.
  • Constant spring tension prevents leakage during low-temperature reactions.
  • Air-operated for rapid, controlled opening and closing.
  • Secondary regulator provides 4 psi safety backup.
  • Wetted parts of PTFE with 316L stainless steel shaft for corrosion resistance.
  • Includes air control panel, two regulators, mounting bracket, tubing, and fittings.
  • Control panel can be mounted outside a hood and positioned using a universal clamp.
Catalog No. 50-273-3548