Learn More
Electron Microscopy Sciences Anti-Vibration Platform Air Preparation Unit
SDP
Supplier:  Electron Microscopy Sciences 6620
Encompass_Preferred
Used if compressed air is used which may be damp or contain small particles. The unit will ensure the air supply is properly cleaned and dry before entering the platforms.
Catalog No. 50-344-69