Learn More
Med Vet International Dynarex Nasopharyngeal Airway, 34 Fr, 8.5 mm, 10/Box
Supplier:  Med Vet International 36282
Encompass_Preferred
Dynarex Nasopharyngeal Airway Kits include an assortment of airways in multiple sizes to accommodate patients’ specific needs, and packets of lubrication jelly for added convenience. Each kit comes with either 6 or 9 airways for ultimate versatility. The Dynarex Nasopharyngeal Airway is designed for nasal insertion to help create and maintain an open and secure airway. The Dynarex Nasopharyngeal Airway features a fixed flange end for safer insertion and internal striations for optimal suctioning efficiency. Dynarex Nasopharyngeal Airway Kits include an assortment of airways in multiple sizes for optimal versatility and packets of lubrication jelly for added convenience. 
Catalog No. 50-283-0800