Learn More
AdipoGen Ajmalicine,Chemical. CAS: 483-04-5
SDP
Supplier:  AdipoGen CDXA0214M250
Encompass_Preferred
Ajmalicine, AdipoGen Life Sciences. Chemical. CAS: 483-04-5. Formula: C21H24N2O3. MW: 352.43. Synthetic. Alkaloid used to study its effects as an antagonist of adrenergic and nicotinic receptors. 483-04-5
Catalog No. 50-117-7200