Learn More
Medchemexpress LLC AJS1669 free acid | 1853130-05-8 | 99.2% | 1 ML

Supplier: Medchemexpress LLC HY12087210MM 1ML
AJS1669 free acid is a small-molecule glycogen synthase (GS) activator used in preclinical research. It has been reported to improve glucose metabolism and reduce body fat mass in mice. The product is supplied as the free acid and is available as powder and as a ready-to-use 10 mM solution in DMSO.
- Potent glycogen synthase activator for metabolic research.
- Demonstrated improvement in glucose metabolism and reduction in body fat in mice.
- Available as powder (5-500 mg) or as 10 mM solution in DMSO (1 mL).
- High purity (99.15%), suitable for in vitro and in vivo studies.
- Stable under recommended storage conditions for powder and stock solutions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.