Learn More
Akina Inc Poly(lactide-co-glycolide)-b-poly(ethylene glycol)-b-poly(lactide-co-glycolide) (per g)
SDP
Supplier:  Akina Inc AK0121G
Encompass
PLGA-PEG-PLGA (Mw ~1,000:1,000:1,000 Da) LA:GA 50:50 Poly(lactide-co-glycolide)-b-poly(ethylene glycol)-b-poly(lactide-co-glycolide) (per g)
Catalog No. NC1632817