Learn More
Medchemexpress LLC AK59 | 99.0% | 429.41 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1625635MG
Encompass_Preferred
AK59 is a STING degrader that operates by utilizing HERC4, a hect domain E3 ligase. It is intended for use in the study of autoimmune diseases and cancer.
  • STING degrader
  • Leverages HERC4, a hect domain E3 ligase
  • Applicable in the study of autoimmune diseases
  • Applicable in the study of cancer
  • For research use only
Catalog No. 50-003-93919