Learn More
Medchemexpress LLC AKB-6899 | 1007377-55-0 | 99.9% | 290.25 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY113649
Encompass_Preferred
AKB-6899, a prolyl hydroxylase domain 3 (PHD3) inhibitor, is a selective HIF-2α stabilizer. It also increases soluble form of the VEGF receptor (sVEGFR-1) production from GM-CSF-treated macrophages, and has antitumor and antiangiogenic effects.
  • PHD3 inhibitor
  • Selective HIF-2α stabilizer
  • Increases soluble form of the VEGF receptor (sVEGFR-1) production from GM-CSF-treated macrophages
  • Has antitumor effects
  • Has antiangiogenic effects
Catalog No. 50-004-38364