Learn More
Selleck Chemical LLC Akebia saponin D S5455-1mg
SDP
Supplier:  Selleck Chemical LLC S54551MG
Encompass_Preferred
Akebia saponin D (Asperosaponin VI) the most abundant constituent of the rhizome of Dipsacus asper has been used for the treatment of lower back pain traumatic hematoma and bone fractures
Catalog No. 50-311-1529