Learn More
ABclonal Technology AKR1A1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A13577500UL
Encompass_Preferred
This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. This member, also known as aldehyde reductase, is involved in the reduction of biogenic and xenobiotic aldehydes and is present in virtually every tissue. Multiple alternatively spliced transcript variants of this gene exist, all encoding the same protein.
Catalog No. 50-001-15668