Learn More
Medchemexpress LLC HY-107379 10mg Medchemexpress, AKR1C3-IN-1 CAS:327092-81-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10737910MG
Encompass_Preferred
Medchemexpress, HY-107379 10mg AKR1C3-IN-1 CAS:327092-81-9 AKR1C3-IN-1 is a potent, highly selective inhibitor of AKR1C3, with an IC50 of 13 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8392