Learn More
Medchemexpress LLC AKR1C3-IN-4 | 1284180-11-5 | 99.8% | 281.23 | 1 ML

Supplier: Medchemexpress LLC HY13855710MM/1ML
AKR1C3-IN-4 is a potent and selective aldo-keto reductase 1C3 (AKR1C3) inhibitor with an IC50 of 0.56 μM. It also has an IC50 of 15.1 μM for AKR1C2. This compound has potential for research related to castrate resistant prostate cancer (CRPC).
- Potent and selective AKR1C3 inhibitor
- IC50 of 0.56 μM for AKR1C3
- IC50 of 15.1 μM for AKR1C2
- Potential for castrate resistant prostate cancer (CRPC) research
- Purity of 99.8%
- Molecular weight of 281.23
- White to yellow solid appearance
- Store at 4°C, protected from light (solid form)
- Store at -80°C for 6 months or -20°C for 1 month in solvent, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.