Learn More
Medchemexpress LLC Akr1c3-in-4 | 1284180-11-5 | 99.8% | 281.23 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13855750MG
Encompass_Preferred
AKR1C3-IN-4 is a potent and selective aldo-keto reductase 1C3 (AKR1C3) inhibitor with an IC50 of 0.56 μM.
  • Has the potential for castrate resistant prostate cancer (CRPC) research.
Catalog No. 50-003-49356