Learn More
Cayman Chemical Akt InhIbItr XI 10mg
SDP
Supplier:  Cayman Chemical 1860410 MG
Encompass_Preferred
A cell-permeable, copper-containing 3-formylchromone derivative that inhibits Akt in an array of cancer cells (IC50s = 10-34 µM); also causes NF-κB inactivation in an orthotopic pancreatic tumor model using COLO 357 cells
Catalog No. 50-297-9664