Learn More
Sino Biological Peptide Substrates: AKT (SGK) Substrate
Supplier:  Sino Biological A0858
Encompass_Preferred
The AKT (SGK) Substrate peptide sequence (RPRAATF) is based on the N-terminus of GSK3.
Catalog No. 50-256-9849