Learn More
Meso Scale Discovery Akt Signaling WCL
SDP
Supplier:  Meso Scale Discovery K15115D2
Encompass
The Meso Scale Discovery Akt Signaling Whole Cell Lysate Kit (K15115D) is designed to monitor Akt phosphorylation and activation in cells The kit includes a 96-well plate coated with capture antibodies for total and phosphorylated Akt (Ser473 and Thr308) It also includes detection antibodies and buffers needed to perform an ELISA-based assay to measure Akt phosphorylation The kit allows for high-throughput analysis of Akt signaling with as little as 2 ug of cell lysate per well Results are quantified on the MSD instrument platform and data is reported as electrochemiluminescence signal This kit enables robust and reproducible analysis of Akt activation and phosphorylation
Catalog No. 50-000-82418
Add to Cart
Request a Quote