Learn More
Medchemexpress LLC Akt1-IN-1 | 99.7% | 637.74 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY146459
Encompass_Preferred
Akt1-IN-1 is a potent and selective Akt1 inhibitor with an IC50 value of 18.79 nM in MIA Paca-2 cells. It does not exhibit obvious teratogenicity, hepatotoxicity, and cardiotoxicity. This compound can be used for researching anticancer. It is for research use only and not sold to patients.
  • Potent and selective Akt1 inhibitor
  • IC50 value of 18.79 nM in MIA Paca-2 cells
  • Does not exhibit obvious teratogenicity
  • Does not exhibit obvious hepatotoxicity
  • Does not exhibit obvious cardiotoxicity
  • Suitable for researching anticancer
Catalog No. 50-003-63173