Learn More
ABclonal Technology AKT2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2400920UL
Encompass_Preferred
This gene is a putative oncogene encoding a protein belonging to a subfamily of serine threonine kinases containing SH2-like (Src homology 2-like) domains which is involved in signaling pathways The gene serves as an oncogene in the tumorigenesis of cancer cells For example its overexpression contributes to the malignant phenotype of a subset of human ductal pancreatic cancers The encoded protein is a general protein kinase capable of phophorylating several known proteins and has also been implicated in insulin signaling
Catalog No. 50-000-07341