Learn More
Insphero Inc Akura™ 3D SureKit - powered by InSphero
SDP
Supplier:  Insphero Inc KT0500201
Encompass
The Akura™ 3D SureKit provides a validated all-in-one solution to generate and culture organotypic 3D spheroids from primary human hepatocytes.The kit contains frozen primary human hepatocytes and specialized media to aggregate the cells and culture formed spheroids. 3D optimized Akura™ 96 Spheroid Microplates are included to allow for homogenous aggregation and loss-free media exchanges. Reference characterization data and dose-response curves for model DILI compounds are provided as benchmark data for your in-house experiments.Components:- 3 x Akura™ 96 Spheroid Microplates- 1 x Akura™ Tilting Stand- Primary human hepatocytes, cryopreserved (> 1 million per vial)- 40 ml Aggregation base medium - 12.5 ml Aggregation supplement - 200 ml Maintenance base medium - 10 ml Maintenance supplement
Catalog No. NC2216389