Learn More
TARGETMOL CHEMICALS INC AL 082D06 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T174925MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. AL 082D06 (D-06), a selective nonsteroidal glucocorticoid receptor (GR) antagonist (Ki 210 nM), exhibits outstanding selectivity against AR, ER, MR and PR(Ki > 10 uM). Purity ≥95%
Catalog No. 50-287-6884