Learn More
Medchemexpress LLC AL 8697 | 1057394-06-5 | 99.8% | 402.41 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY108645
Encompass_Preferred
AL 8697 is a specific and orally active p38α MAPK inhibitor with an IC50 of 6 nM. It exhibits 14-fold greater inhibition of p38α compared to p38β (IC50=82 nM) and demonstrates 300-fold selectivity for p38α over a panel of 91 kinases. This compound also possesses anti-inflammatory activity and is for research use only, not for sale to patients.
  • Specific and orally active p38α MAPK inhibitor
  • High selectivity for p38α over p38β and other kinases
  • Anti-inflammatory activity
  • For research use only
Catalog No. 50-003-47491