Learn More
TARGETMOL CHEMICALS INC AL 8697 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T102775MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AL 8697 is a selective p38(alpha) MAPK inhibitor (IC50 = 6 nM) with 14-fold selectivity over p38(beta) (IC50 = 82 nM) and 300-fold selectivity over a panel of 91 kinases. AL 8697 has anti-inflammatory activity. purity: 99%
Catalog No. 50-258-1740