Learn More
Medchemexpress LLC Ala-D-γ-Glu-Lys-D-Ala-D-Ala | 2614-55-3 | 99.8% | 488.54 | 25 MG

Supplier: Medchemexpress LLC 25MGHYP5919
Ala-D-γ-Glu-Lys-D-Ala-D-Ala is a pentapeptide that functions as the tail of the peptidoglycan precursor UDPMurNAc-l-Ala-γ-d-Glu-l-Lys(Gly)(5)-d-Ala-d-Ala. It is intended for laboratory chemicals and the manufacture of substances, strictly for research use only.
- CAS number: 2614-55-3
- Molecular formula: C20H36N6O8
- Molecular weight: 488.54
- Purity: 99.8%
- Appearance: Solid
- Recommended storage for powder: -80°C for 2 years, -20°C for 1 year
- Recommended storage for in solvent: -80°C for 6 months, -20°C for 1 month
- Causes skin irritation (H315)
- Causes serious eye irritation (H319)
- Solubility in H₂O: ≥ 100 mg/mL (204.69 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.