Learn More
Medchemexpress LLC Ala-Phe-Pro-pNA TFA | 99.9% | 453.49 | 5 MG

Supplier: Medchemexpress LLC HYP4464A5MG
Ala-Phe-Pro-pNA TFA is a chromogenic substrate of tripeptidyl peptidase and can be used to test tripeptidyl peptidase activity.
- Purity: 99.9%
- Molecular weight: 453.49 (free base)
- Chemical formula: C23H27N5O5.xC2HF3O2
- Appearance: Solid
- Color: White to off-white
- Sequence: Ala-Phe-{Pro-pNA}
- Sequence shortening: AF-{Pro-pNA}
- In vitro solubility: DMSO: ≥ 100 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.