Learn More
Medchemexpress LLC [Ala17]-MCH | 359784-84-2 | 98.3% | 2271.71 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP120410MG
Encompass_Preferred
[Ala17]-MCH is a synthetic peptide, acting as a selective ligand for the MCHR1 receptor (Ki=0.16 nM) with significantly lower affinity for MCHR2 (Ki=34 nM). A europium-chelate labeled version of [Ala17]-MCH demonstrates high affinity for MCHR1 (Kd=0.37 nM) and minimal binding affinity for MCHR2. This compound is supplied as a white to off-white solid.
  • Selective ligand for MCHR1
  • High affinity for MCHR1 (Ki=0.16 nM)
  • Minimal binding affinity for MCHR2 (Ki=34 nM)
  • Soluble in DMSO at 25 mg/mL
  • Multiple in vivo dissolution protocols available
Catalog No. 50-004-22329