Learn More
Medchemexpress LLC Alacepril 10mM 1mL | 74258-86-9 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY10731810MM 1ML
Encompass_Preferred
Alacepril (Cetapril) is an orally active angiotensin converting enzyme (ACE) inhibitor with long lasting antihypertensive effect[1
Catalog No. 50-002-41518