Learn More
Medchemexpress LLC Alacepril | 74258-86-9 | 98.0% | 406.50 g·mol⁻¹ | C20H26N2O5S | 50 MG

Supplier: Medchemexpress LLC HY10731850MG
Alacepril is an orally active angiotensin-converting enzyme (ACE) inhibitor used in research as a reference antihypertensive compound. Supplied as a solid reagent, it is intended for pharmacology and biochemical assays studying ACE inhibition and cardiovascular effects.
- Orally active ACE inhibitor chemical.
- Long-lasting antihypertensive effect.
- High purity (98.0%) suitable for research applications.
- Supplied as a solid for ease of handling and storage.
- Molecular weight 406.50 g·mol⁻¹; molecular formula C20H26N2O5S.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.