Learn More
Medchemexpress LLC Alagebrium chloride | 341028-37-3 | MFCD09027398 | 99.8% | 267.78 g·mol⁻¹ | C13H14ClNOS | 1 ML
SDP
Supplier:  Medchemexpress LLC HY106024B10MM 1ML
Encompass_Preferred
Alagebrium chloride (ALT711) is an advanced glycation end product (AGE) inhibitor used in research to disrupt and reverse glucose-derived cross-links. It is supplied as a 10 mM stock solution in DMSO or as a solid for reconstitution and is intended for laboratory research use only.
  • Inhibits advanced glycation end product (AGE) formation and breaks glucose cross-links.
  • Provided as a 10 mM solution in DMSO or as a solid for reconstitution.
  • High purity suitable for analytical and biological assays (reported ≈99.8%).
  • Soluble in DMSO and water; in vivo formulation guidance available in datasheet.
  • Stable under recommended storage; in solvent: -80°C (2 years) or -20°C (1 year).
Catalog No. 50-002-43168