Learn More
Medchemexpress LLC Alamifovir | 193681-12-8 | 95.0% | 1 MG

Supplier: Medchemexpress LLC HY1196911MG
Alamifovir is a purine nucleotide analogue prodrug with potent activity against wild-type and lamivudine-resistant hepatitis B virus (HBV). It is intended for research use in antiviral and in vitro pharmacology studies.
- Shows potent activity against HBV replication and sustained antiviral effect.
- Suitable for in vitro antiviral and resistance studies.
- Purity 95.0%.
- Molecular formula C19H20F6N5O5PS; molecular weight 575.42 g/mol.
- Supplied in small research quantities for assay work.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.