Learn More
Medchemexpress LLC β-Alanine-15N | 204451-53-6 | 98.0% | 90.09 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0230S1
Encompass_Preferred
β-Alanine-15N is the 15N-labeled β-Alanine, a non-essential amino acid. It is metabolized into carnosine, which functions as an intracellular buffer. This compound is suitable for various analytical applications.
  • Used as a tracer for biological pathways.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes, beneficial for drug development quantitation.
Catalog No. 50-004-29907