Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-alanine-d4 (2-carboxyethylamine-d4) | 116173-67-2 | 99.9% | 93.12 | C3H3D4NO2 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN0230S210MG

Encompass_Preferred

β-Alanine-d4 (2-carboxyethylamine-d4) is the deuterium-labeled form of β-alanine used as an isotopic standard and tracer in analytical and biochemical applications. It is supplied as a high-purity solid suitable for mass spectrometry, NMR, and metabolic tracing studies.

  • Deuterium-labeled (d4) β-alanine for isotopic tracing and standards.
  • High purity (99.9%) for accurate analytical results.
  • Molecular weight 93.12 and formula C3H3D4NO2.
  • CAS number 116173-67-2 for unambiguous identification.
  • Store protected from light; recommended storage at 4°C; in solvent: -80°C up to 6 months, -20°C up to 1 month.

Catalog No. 50-002-95632


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.